Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZNF385C | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001242704.1 | NP_001229633.1 | 8 | 1528 | ||
XM_006721751.3 | XP_006721814.1 | 8 | 1636 | |||
XM_011524479.2 | XP_011522781.1 | 9 | 2332 | |||
XM_011524482.2 | XP_011522784.1 | 9 | 2329 | |||
XM_011524483.2 | XP_011522785.1 | 9 | 2318 | |||
XM_017024314.1 | XP_016879803.1 | 10 | 2461 | |||
XM_017024315.1 | XP_016879804.1 | 10 | 2296 | |||
XM_017024316.1 | XP_016879805.1 | 10 | 2458 | |||
XM_017024317.1 | XP_016879806.1 | 10 | 2447 | |||
XM_017024318.1 | XP_016879807.1 | 10 | 2445 | |||
XM_017024319.1 | XP_016879808.1 | 9 | 2315 | |||
XM_017024320.1 | XP_016879809.1 | 9 | 1723 | |||
GenBank mRNA | BC080613.1 | - | 4 | 921 | 5922386 | |
CN264203.1 | - | 1 | 212 | |||
H06009.1 | - | 1 | 300 |