Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SAMD14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001257359.1 | NP_001244288.1 | 8 | 1306 | ||
NM_174920.3 | NP_777580.1 | 9 | 1390 | |||
XM_011524490.2 | XP_011522792.1 | 5 | 492 | |||
XM_017024322.1 | XP_016879811.1 | 7 | 1690 | |||
XM_017024323.1 | XP_016879812.1 | 9 | 1473 | |||
XM_017024324.1 | XP_016879813.1 | 9 | 2245 | |||
GenBank mRNA | AF035306.1 | - | 2 | 178 | ||
AK054843.1 | - | 1 | 824 | |||
AK094209.1 | - | 8 | 1237 | |||
AK095809.1 | - | 5 | 607 | |||
AK225767.1 | - | 8 | 1237 | |||
AK291482.1 | - | 8 | 1233 | |||
AY148482.1 | - | 9 | 1311 | |||
BC029872.1 | - | 4 | 418 | |||
BC047081.1 | - | 7 | 960 | |||
BC136485.1 | - | 8 | 1219 | 9020474 | ||
BC141822.1 | - | 8 | 1246 | 40148162 | ||
BC142728.1 | - | 8 | 1203 | 40148957 |