Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SPNS3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001320449.1 | NP_001307378.1 | 1 | 227 | ||
NM_182538.4 | NP_872344.3 | 1 | 227 | |||
XM_011523723.1 | XP_011522025.1 | 1 | 211 | |||
XM_011523724.1 | XP_011522026.1 | 1 | 211 | |||
XM_011523725.1 | XP_011522027.1 | 1 | 211 | |||
GenBank mRNA | AK172832.1 | - | 1 | 227 | ||
BC023646.2 | - | 1 | 179 | 5087991 |