Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZSCAN4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_152677.2 | NP_689890.1 | 5 | 1358 | ||
XM_011526607.2 | XP_011524909.1 | 7 | 1539 | |||
XM_017026458.1 | XP_016881947.1 | 5 | 1758 | |||
GenBank mRNA | AK092424.1 | - | 5 | 1357 | ||
BC101738.1 | - | 3 | 749 | 8069244 | ||
BC101740.1 | - | 3 | 749 | 8069246 |