Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CDMO-CRO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other PRIMPOL SIRNA Assays ›
Gene Symbol:
PRIMPOL
Gene Name:
primase and DNA directed polymerase
Gene Aliases:
CCDC111, MYP22
Chromosome Location:
Chr.4: 184649613 - 184694959 on Build GRCh38
UniGene ID:
Hs.481307
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
s47416
Purification
HPLC
Standard
HPLC-IVR
Size
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
Availability
Inventoried
Catalog #
4392420
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: PRIMPOL
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001300767.1
NP_001287696.1
10
1470
NM_001300768.1
NP_001287697.1
11
1706
NM_152683.3
NP_689896.1
11
1709
XM_011531719.1
XP_011530021.1
11
1586
XM_011531720.1
XP_011530022.1
11
1624
XM_011531721.2
XP_011530023.1
11
1583
XM_011531723.1
XP_011530025.1
10
1411
XM_011531724.2
XP_011530026.1
9
1368
XM_011531725.1
XP_011530027.1
9
1282
XM_011531726.2
XP_011530028.1
8
1129
XM_011531729.1
XP_011530031.1
10
1415
XM_011531730.2
XP_011530032.1
10
1430
XM_017007864.1
XP_016863353.1
11
1621
XM_017007865.1
XP_016863354.1
11
1624
XM_017007866.1
XP_016863355.1
11
1621
XM_017007867.1
XP_016863356.1
11
1586
XM_017007868.1
XP_016863357.1
11
1586
XM_017007869.1
XP_016863358.1
9
1334
XM_017007870.1
XP_016863359.1
9
1334
XM_017007871.1
XP_016863360.1
9
1367
XM_017007872.1
XP_016863361.1
9
1277
XM_017007873.1
XP_016863362.1
10
1456
XM_017007874.1
XP_016863363.1
9
1275
XM_017007875.1
XP_016863364.1
10
1412
GenBank mRNA
AK057729.1
-
11
1492
BC047467.1
-
10
1416
BC064600.1
-
11
1550
5786393
BX647575.1
-
11
1594
Back To Top
More Information
Panther Classification:
Molecular Function -
DNA metabolism protein
nucleic acid metabolism protein
Gene Ontology Categories:
Function(s)
Process(es)
mitochondrial DNA replication
DNA replication, synthesis of RNA primer
response to UV
translesion synthesis
replication fork processing
chromatin binding
DNA-directed DNA polymerase activity
DNA primase activity
protein binding
manganese ion binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-blue-5c9c5c4749-8tsm6:80/100.66.77.47:80.
git-commit: d22414ba47b22cc02ffc24003f0e179da61511a6
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.21.0-Offline