Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TAPT1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_153365.2 | NP_699196.2 | 6 | 916 | ||
XM_005248139.2 | XP_005248196.1 | 5 | 730 | |||
XM_005248140.3 | XP_005248197.1 | 5 | 680 | |||
XM_011513815.2 | XP_011512117.1 | 5 | 2015 | |||
XM_011513816.2 | XP_011512118.1 | 5 | 724 | |||
XM_011513817.2 | XP_011512119.1 | 3 | 293 | |||
XM_017007876.1 | XP_016863365.1 | 7 | 806 | |||
GenBank mRNA | AK074494.1 | - | 6 | 849 | ||
AK296104.1 | - | 5 | 718 | |||
BC066899.1 | - | 6 | 854 | 4825556 |