Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DNAJC18 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_152686.3 | NP_689899.1 | 3 | 365 | ||
GenBank mRNA | AK001329.1 | - | 1 | 42 | ||
AK308138.1 | - | 3 | 341 | |||
BC030162.2 | - | 3 | 290 | 5020970 | ||
DA497858.1 | - | 3 | 365 |