Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DNAJC18 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_152686.3 | NP_689899.1 | 2 | 133 | ||
GenBank mRNA | AK308138.1 | - | 2 | 109 | ||
BC030162.2 | - | 2 | 58 | 5020970 | ||
DA497858.1 | - | 2 | 133 |