Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FAM153A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_005265889.3 | XP_005265946.1 | 2 | 123 | ||
XM_006714848.3 | XP_006714911.1 | 2 | 145 | |||
XM_011534520.2 | XP_011532822.1 | 2 | 123 | |||
XM_011534528.2 | XP_011532830.1 | 2 | 123 | |||
XM_017009352.1 | XP_016864841.1 | 2 | 123 | |||
XM_017009353.1 | XP_016864842.1 | 2 | 123 | |||
XM_017009354.1 | XP_016864843.1 | 2 | 123 | |||
XM_017009355.1 | XP_016864844.1 | 2 | 123 | |||
XM_017009356.1 | XP_016864845.1 | 2 | 123 | |||
XM_017009357.1 | XP_016864846.1 | 2 | 123 | |||
XM_017009358.1 | XP_016864847.1 | 2 | 123 | |||
XM_017009359.1 | XP_016864848.1 | 2 | 123 | |||
XM_017009360.1 | XP_016864849.1 | 2 | 123 | |||
XM_017009361.1 | XP_016864850.1 | 2 | 123 |
Gene Symbol: FAM153B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BC028606.1 | - | 2 | 176 | 5266126 |