Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: STK32A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001112724.1 | NP_001106195.1 | 3, 4 | 395 | ||
NM_001287740.1 | NP_001274669.1 | 3, 4 | 395 | |||
NM_145001.3 | NP_659438.1 | 3, 4 | 395 | |||
XM_011537576.1 | XP_011535878.1 | 2, 3 | 131 | |||
XM_011537577.2 | XP_011535879.1 | 3, 4 | 448 | |||
XM_011537579.2 | XP_011535881.1 | 3, 4 | 448 | |||
XM_017009213.1 | XP_016864702.1 | 3, 4 | 448 | |||
GenBank mRNA | AK094580.1 | - | 3, 4 | 448 | ||
AK299099.1 | - | 3, 4 | 286 | |||
AK315942.1 | - | 3, 4 | 385 | |||
BC021666.1 | - | 3, 4 | 308 | 3682677 | ||
DA110948.1 | - | 3, 4 | 395 | |||
DR006392.1 | - | 3, 4 | 287 |