Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: EPHB6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001280794.2 | NP_001267723.2 | 8 | 2105 | ||
NM_001280795.2 | NP_001267724.2 | 5 | 1477 | |||
NM_004445.5 | NP_004436.4 | 6 | 1546 | |||
NR_104001.2 | 8 | 1392 | ||||
XM_006715881.2 | XP_006715944.1 | 9 | 2541 | |||
XM_011515879.1 | XP_011514181.1 | 10 | 2580 | |||
XM_011515880.1 | XP_011514182.1 | 9 | 2558 | |||
XM_011515881.2 | XP_011514183.1 | 9 | 2542 | |||
XM_011515882.2 | XP_011514184.1 | 9 | 2544 | |||
GenBank mRNA | BC056159.1 | - | 8 | 1974 | ||
BC110606.1 | - | 5 | 1471 | 40028676 | ||
BC110607.1 | - | 6 | 1540 | 40028687 | ||
D83492.1 | - | 9 | 2117 | |||
EU054308.1 | - | 5 | 1319 |