Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HTRA4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_153692.3 | NP_710159.1 | 3, 4 | 882 | ||
XM_011544431.2 | XP_011542733.1 | 3, 4 | 882 | |||
XM_011544432.1 | XP_011542734.1 | 3, 4 | 882 | |||
GenBank mRNA | AK075205.1 | - | 3, 4 | 853 | ||
AK300197.1 | - | 3, 4 | 761 | |||
BC057765.1 | - | 3, 4 | 814 | 30348811 |