Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: EPHB6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001280794.2 | NP_001267723.2 | 6 | 1004 | ||
NM_001280795.2 | NP_001267724.2 | 3 | 376 | |||
NM_004445.5 | NP_004436.4 | 4 | 445 | |||
NR_104001.2 | 5 | 562 | ||||
XM_006715881.2 | XP_006715944.1 | 7 | 1440 | |||
XM_011515879.1 | XP_011514181.1 | 8 | 1479 | |||
XM_011515880.1 | XP_011514182.1 | 7 | 1457 | |||
XM_011515881.2 | XP_011514183.1 | 7 | 1441 | |||
XM_011515882.2 | XP_011514184.1 | 7 | 1443 | |||
GenBank mRNA | BC056159.1 | - | 6 | 876 | ||
BC110606.1 | - | 3 | 373 | 40028676 | ||
BC110607.1 | - | 4 | 442 | 40028687 | ||
BX410199.2 | - | 3 | 367 | |||
CN283348.1 | - | 1 | 67 | |||
D83492.1 | - | 7 | 1019 | |||
EU054308.1 | - | 3 | 221 |