Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: C9orf72 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001256054.2 | NP_001242983.1 | 2 | 497 | ||
NM_018325.4 | NP_060795.1 | 2 | 402 | |||
NM_145005.6 | NP_659442.2 | 2 | 419 | |||
GenBank mRNA | AK057806.1 | - | 2 | 397 | ||
AK291425.1 | - | 2 | 392 | |||
AL832229.1 | - | 1 | 359 | |||
AL832467.1 | - | 2 | 358 | |||
BC020851.2 | - | 1 | 339 | |||
BC068445.1 | - | 1 | 351 | 5298741 | ||
BG718065.1 | - | 2 | 408 | |||
BG722125.1 | - | 2 | 395 | |||
BQ068108.1 | - | 508 | ||||
JN681271.1 | - | 2 | 475 |