Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CCDC171 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_173550.2 | NP_775821.2 | 25 | 3990 | ||
XM_005251397.2 | XP_005251454.1 | 25 | 3897 | |||
XM_005251398.3 | XP_005251455.1 | 25 | 4136 | |||
XM_005251399.4 | XP_005251456.1 | 26 | 4196 | |||
XM_011517788.2 | XP_011516090.1 | 25 | 4126 | |||
XM_011517791.2 | XP_011516093.1 | 24 | 4004 | |||
XM_011517796.2 | XP_011516098.1 | 23 | 3543 | |||
XM_017014431.1 | XP_016869920.1 | 25 | 4136 | |||
XM_017014432.1 | XP_016869921.1 | 25 | 4136 | |||
XM_017014433.1 | XP_016869922.1 | 25 | 4136 | |||
XM_017014434.1 | XP_016869923.1 | 24 | 4004 | |||
XM_017014436.1 | XP_016869925.1 | 25 | 4136 | |||
XM_017014437.1 | XP_016869926.1 | 25 | 4136 | |||
XM_017014438.1 | XP_016869927.1 | 25 | 4135 | |||
XM_017014439.1 | XP_016869928.1 | 26 | 4298 | |||
XM_017014440.1 | XP_016869929.1 | 23 | 3557 | |||
XM_017014443.1 | XP_016869932.1 | 24 | 3618 | |||
XM_017014444.1 | XP_016869933.1 | 14 | 2337 | |||
GenBank mRNA | AF440401.1 | - | 5 | 471 | ||
AK128581.1 | - | 13 | 2253 | |||
AL832443.1 | - | 2 | 210 | |||
AY422473.1 | - | 25 | 3990 | |||
BC064849.1 | - | 12 | 2196 | |||
BC144214.1 | - | 24 | 3791 | 9052734 | ||
BC150600.1 | - | 24 | 3791 | 9056970 | ||
CR936775.1 | - | 26 | 4268 |