Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SUSD3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001287005.1 | NP_001273934.1 | 6 | 1160 | ||
NM_001287006.1 | NP_001273935.1 | 4 | 914 | |||
NM_001287007.1 | NP_001273936.1 | 3 | 725 | |||
NM_001287008.1 | NP_001273937.1 | 4 | 857 | |||
NM_145006.3 | NP_659443.1 | 5 | 1046 | |||
XM_011518358.1 | XP_011516660.1 | 8 | 2240 | |||
XM_011518359.1 | XP_011516661.1 | 7 | 2190 | |||
XM_011518361.2 | XP_011516663.1 | 8 | 1302 | |||
XM_011518362.1 | XP_011516664.1 | 6 | 2076 | |||
XM_017014448.1 | XP_016869937.1 | 6 | 1347 | |||
XM_017014449.1 | XP_016869938.1 | 5 | 1228 | |||
XM_017014450.1 | XP_016869939.1 | 7 | 1187 | |||
XM_017014451.1 | XP_016869940.1 | 5 | 1796 | |||
GenBank mRNA | AK128289.1 | - | 3 | 3014 | ||
AY358190.1 | - | 6 | 1088 | |||
BC014601.2 | - | 5 | 1027 | 4821517 | ||
BC041834.1 | - | 3 | 2940 | 5268318 | ||
BM458621.1 | - | 3 | 603 |