Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: VMA21 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001017980.3 | NP_001017980.1 | 2 | 271 | ||
XM_011531125.2 | XP_011529427.1 | 2 | 597 | |||
GenBank mRNA | AK096835.1 | - | 2 | 250 | ||
AL833596.1 | - | 2 | 224 | |||
BC103701.1 | - | 2 | 228 | 40023465 | ||
BC103702.1 | - | 2 | 228 | 40023460 | ||
BC105693.1 | - | 2 | 228 | 40023462 | ||
BC105694.1 | - | 2 | 228 | 40023459 | ||
BC110800.1 | - | 2 | 184 | 6044374 | ||
DA224893.1 | - | 2 | 271 |