Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NLRP11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001297743.1 | NP_001284672.1 | 7 | 2464 | ||
NM_145007.3 | NP_659444.2 | 10 | 3314 | |||
GenBank mRNA | AK090621.1 | - | 5 | 1352 | ||
AK129784.1 | - | 5 | 1379 | |||
AY095145.1 | - | 8 | 2702 | |||
AY154466.1 | - | 7 | 2603 | |||
AY168967.1 | - | 8 | 2691 | |||
AY168968.1 | - | 7 | 2363 | |||
AY226382.1 | - | 7 | 2603 | |||
BC034730.1 | - | 8 | 2691 | 4341197 |