Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AMER3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001105193.1 | NP_001098663.1 | 2 | 2055 | ||
NM_001105194.1 | NP_001098664.1 | 2 | 1991 | |||
NM_001105195.1 | NP_001098665.1 | 2 | 1988 | |||
NM_152698.2 | NP_689911.2 | 2 | 2071 | |||
XM_011510793.2 | XP_011509095.1 | 2 | 3097 | |||
GenBank mRNA | AK095696.1 | - | 2 | 2035 | ||
AK289525.1 | - | 2 | 2140 | |||
AK291424.1 | - | 2 | 2035 | |||
BC113445.1 | - | 2 | 1948 | 8322497 | ||
BC113447.1 | - | 2 | 1948 | 8322499 | ||
BC143806.1 | - | 2 | 1945 | 9052321 |