Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LVRN | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_173800.4 | NP_776161.3 | 18 | 2874 | ||
GenBank mRNA | AK075131.1 | - | 13 | 1409 | ||
AK291655.1 | - | 18 | 2888 | |||
AY560010.1 | - | 18 | 2762 | |||
BC036440.1 | - | 13 | 1753 | |||
BC045809.1 | - | 12 | 1471 | |||
BC060869.1 | - | 19 | 2801 | 30346049 | ||
BC068560.1 | - | 17 | 2720 | 30341970 | ||
BC070028.1 | - | 13 | 1871 | |||
BC094716.1 | - | 13 | 1871 | |||
BC109022.1 | - | 18 | 2738 | 40008501 | ||
BC109023.1 | - | 18 | 2738 | 40008502 |