Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CDMO-CRO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other SLC36A1 SIRNA Assays ›
Gene Symbol:
SLC36A1
Gene Name:
solute carrier family 36 member 1
Gene Aliases:
Dct1, LYAAT1, PAT1, TRAMD3
Chromosome Location:
Chr.5: 151411344 - 151556318 on Build GRCh38
UniGene ID:
Hs.269004
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
s47577
Purification
HPLC
Standard
HPLC-IVR
Size
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
Availability
Inventoried
Catalog #
4392420
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: SLC36A1
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001308150.1
NP_001295079.1
7
923
NM_001308151.1
NP_001295080.1
7
893
NM_078483.3
NP_510968.2
7
923
XM_005268386.1
XP_005268443.1
7
893
XM_006714759.3
XP_006714822.1
7
972
XM_011537580.2
XP_011535882.1
7
1613
XM_011537581.1
XP_011535883.1
8
1041
XM_011537583.2
XP_011535885.1
8
5478
XM_011537584.2
XP_011535886.1
7
1132
XM_011537585.1
XP_011535887.1
8
988
XM_011537586.2
XP_011535888.1
7
1736
XM_011537587.2
XP_011535889.1
7
992
XM_011537588.2
XP_011535890.1
7
836
XM_011537589.2
XP_011535891.1
7
1744
XM_011537590.1
XP_011535892.1
7
751
XM_011537591.1
XP_011535893.1
7
726
XM_011537592.2
XP_011535894.1
7
730
XM_011537593.2
XP_011535895.1
8
986
XM_011537594.1
XP_011535896.1
9
1100
XM_011537595.2
XP_011535897.2
7
1619
XM_011537596.2
XP_011535898.2
6
705
XM_017009216.1
XP_016864705.1
7
819
XM_017009217.1
XP_016864706.1
7
923
XM_017009218.1
XP_016864707.1
6
705
XM_017009219.1
XP_016864708.1
7
973
XM_017009220.1
XP_016864709.1
7
893
GenBank mRNA
AF516142.1
-
6
715
AK057340.1
-
7
893
AK309323.1
-
7
895
AY162213.1
-
7
828
AY227111.1
-
8
843
AY227112.1
-
1
211
AY227113.1
-
7
799
BC136437.1
-
7
794
9020426
BC136438.1
-
7
794
9020427
BX537963.1
-
7
920
GU296670.1
-
6
700
Back To Top
More Information
Panther Classification:
Molecular Function -
amino acid transporter
Gene Ontology Categories:
Function(s)
Process(es)
ion transport
amino acid transport
L-alanine transport
glycine transport
proline transport
proline transmembrane transport
hydrogen ion transmembrane transport
hydrogen:amino acid symporter activity
hydrogen ion transmembrane transporter activity
amino acid transmembrane transporter activity
L-alanine transmembrane transporter activity
glycine transmembrane transporter activity
L-proline transmembrane transporter activity
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-blue-5c9c5c4749-97l6p:80/100.66.76.85:80.
git-commit: d22414ba47b22cc02ffc24003f0e179da61511a6
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.21.0-Offline