Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DHRSX | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145177.2 | NP_660160.2 | 5 | 479 | ||
GenBank mRNA | AJ293620.1 | - | 5 | 439 | ||
AL137300.1 | - | 1 | 53 | |||
AY358849.1 | - | 5 | 441 | |||
BC019696.2 | - | 5 | 432 | |||
BC032340.1 | - | 5 | 443 | 5192975 | ||
BG703185.1 | - | 5 | 536 | |||
BM794858.1 | - | 2 | 159 |