Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: USP12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_182488.3 | NP_872294.2 | 4 | 717 | ||
XM_005266282.2 | XP_005266339.1 | 4 | 631 | |||
XM_011534956.2 | XP_011533258.1 | 4 | 468 | |||
XM_017020416.1 | XP_016875905.1 | 3 | 388 | |||
GenBank mRNA | AF022789.1 | - | 4 | 567 | ||
BC026072.1 | - | 4 | 718 | 4830251 |