Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: USP12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_182488.3 | NP_872294.2 | 7 | 1112 | ||
XM_005266282.2 | XP_005266339.1 | 7 | 1026 | |||
XM_011534956.2 | XP_011533258.1 | 7 | 863 | |||
XM_017020416.1 | XP_016875905.1 | 6 | 783 | |||
GenBank mRNA | AF022789.1 | - | 7 | 962 | ||
AK289685.1 | - | 7 | 1026 | |||
BC026072.1 | - | 7 | 1113 | 4830251 |