Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FOLH1B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_153696.2 | NP_710163.1 | 12, 13 | 1582 | ||
XM_017017319.1 | XP_016872808.1 | 17, 18 | 1935 | |||
GenBank mRNA | AF261715.1 | - | 12, 13 | 1582 | ||
AY544126.1 | - | 11, 12 | 1081 |