Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FOLH1B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_153696.2 | NP_710163.1 | 4 | 661 | ||
XM_017017319.1 | XP_016872808.1 | 9 | 1014 | |||
GenBank mRNA | AF261715.1 | - | 4 | 661 | ||
AY544126.1 | - | 3 | 160 |
Gene Symbol: FOLH1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BC108719.1 | - | 7 | 824 |