Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ARMC3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001282745.1 | NP_001269674.1 | 11 | 1307 | ||
NM_001282746.1 | NP_001269675.1 | 11 | 1307 | |||
NM_001282747.1 | NP_001269676.1 | 8 | 818 | |||
NM_173081.4 | NP_775104.2 | 11 | 1307 | |||
XM_005252380.2 | XP_005252437.1 | 10 | 1095 | |||
XM_005252381.2 | XP_005252438.1 | 10 | 1128 | |||
XM_011519350.2 | XP_011517652.1 | 11 | 1290 | |||
XM_011519351.2 | XP_011517653.1 | 2 | 137 | |||
XM_017015831.1 | XP_016871320.1 | 10 | 1095 | |||
XM_017015832.1 | XP_016871321.1 | 11 | 1290 | |||
XM_017015833.1 | XP_016871322.1 | 10 | 1095 | |||
GenBank mRNA | AK057389.1 | - | 11 | 1250 | ||
AK302104.1 | - | 8 | 765 | |||
AK302985.1 | - | 11 | 1294 | |||
AY436643.1 | - | 11 | 1268 | |||
BC039312.1 | - | 11 | 1300 | 5296302 |