Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TYSND1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001040273.2 | NP_001035363.1 | 2 | 1792 | ||
NM_173555.3 | NP_775826.2 | 4 | 2109 | |||
NR_073580.1 | 2 | 560 | ||||
NR_073581.1 | 4 | 997 | ||||
NR_073582.1 | 4 | 877 | ||||
NR_073591.1 | 4 | 788 | ||||
NR_073592.1 | 2 | 680 | ||||
NR_073593.1 | 4 | 780 | ||||
NR_073594.1 | 4 | 900 | ||||
NR_073595.1 | 4 | 1033 | ||||
GenBank mRNA | AK074968.1 | - | 4 | 935 | ||
AK097697.1 | - | 3 | 1652 | |||
BC016840.1 | - | 4 | 1563 | |||
BC030242.1 | - | 2 | 1708 | 5176098 | ||
BC047424.1 | - | 2 | 560 | |||
BC111501.1 | - | 4 | 2048 | 5732356 |