Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: EPS15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001159969.1 | NP_001153441.1 | 4 | 779 | ||
NM_001981.2 | NP_001972.1 | 16 | 1648 | |||
XM_005270618.2 | XP_005270675.1 | 15 | 1589 | |||
XM_017000615.1 | XP_016856104.1 | 16 | 1790 | |||
XM_017000619.1 | XP_016856108.1 | 17 | 1672 | |||
XM_017000620.1 | XP_016856109.1 | 16 | 1839 | |||
GenBank mRNA | AK090936.1 | - | 4 | 505 | ||
AK294297.1 | - | 5 | 628 | |||
AK313396.1 | - | 16 | 1597 | |||
BX647676.1 | - | 4 | 760 | |||
CD657119.1 | - | 3 | 423 | |||
DQ367924.1 | - | 16 | 1556 | |||
U07707.1 | - | 16 | 1577 | |||
Z29064.1 | - | 16 | 1648 |