Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: EPS15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001981.2 | NP_001972.1 | 10 | 876 | ||
XM_005270618.2 | XP_005270675.1 | 9 | 817 | |||
XM_017000615.1 | XP_016856104.1 | 10 | 1018 | |||
XM_017000616.1 | XP_016856105.1 | 10 | 1018 | |||
XM_017000617.1 | XP_016856106.1 | 10 | 907 | |||
XM_017000618.1 | XP_016856107.1 | 9 | 817 | |||
XM_017000619.1 | XP_016856108.1 | 11 | 900 | |||
XM_017000620.1 | XP_016856109.1 | 10 | 1067 | |||
GenBank mRNA | AK313396.1 | - | 10 | 825 | ||
BC054006.1 | - | 11 | 794 | 6164633 | ||
DQ367924.1 | - | 10 | 784 | |||
U07707.1 | - | 10 | 805 | |||
Z29064.1 | - | 10 | 876 |