Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TMEM136 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001198670.1 | NP_001185599.1 | 3 | 1231 | ||
NM_001198671.1 | NP_001185600.1 | 3 | 912 | |||
NM_001198672.1 | NP_001185601.1 | 3 | 901 | |||
NM_001198673.1 | NP_001185602.1 | 3 | 887 | |||
NM_001198674.1 | NP_001185603.1 | 4 | 555 | |||
NM_001198675.1 | NP_001185604.1 | 2 | 712 | |||
NM_174926.2 | NP_777586.1 | 4 | 874 | |||
XM_017017327.1 | XP_016872816.1 | 3 | 3458 | |||
GenBank mRNA | AK096207.1 | - | 2 | 546 | ||
AK128040.1 | - | 3 | 916 | |||
AK289680.1 | - | 3 | 729 | |||
AK294713.1 | - | 4 | 383 | |||
AK304086.1 | - | 3 | 1064 | |||
BC015232.1 | - | 4 | 696 | 3900115 |