Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GLYATL2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145016.3 | NP_659453.3 | 5 | 529 | ||
XM_011544809.2 | XP_011543111.1 | 5 | 714 | |||
XM_017017337.1 | XP_016872826.1 | 6 | 664 | |||
XM_017017338.1 | XP_016872827.1 | 5 | 1111 | |||
GenBank mRNA | AB207211.1 | - | 4 | 398 | ||
AF426250.1 | - | 5 | 789 | |||
BC016789.1 | - | 6 | 673 | 4091916 | ||
BC021682.1 | - | 5 | 516 | 4079236 |