Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DDIAS | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145018.3 | NP_659455.3 | 6 | 3029 | ||
XM_011544835.1 | XP_011543137.1 | 5 | 2891 | |||
XM_011544836.2 | XP_011543138.1 | 5 | 2846 | |||
XM_011544837.1 | XP_011543139.1 | 4 | 2741 | |||
XM_011544838.2 | XP_011543140.1 | 5 | 2875 | |||
XM_017017352.1 | XP_016872841.1 | 4 | 2746 | |||
XM_017017353.1 | XP_016872842.1 | 4 | 2713 | |||
GenBank mRNA | AK297366.1 | - | 4 | 2714 | ||
BC039268.1 | - | 6 | 2997 | 5296465 |