Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LRTOMT | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001205138.3 | NP_001192067.1 | 5 | 1079 | ||
NM_001271471.2 | NP_001258400.1 | 5 | 1498 | |||
NM_001318803.1 | NP_001305732.1 | 6 | 1173 | |||
NM_145309.5 | NP_660352.1 | 6 | 1216 | |||
NR_026886.3 | 7 | 1396 | ||||
NR_134858.1 | 4 | 995 | ||||
XM_006718473.3 | XP_006718536.1 | 6 | 1442 | |||
XM_006718474.3 | XP_006718537.1 | 5 | 1481 | |||
XM_011544847.2 | XP_011543149.1 | 6 | 1670 | |||
XM_011544848.2 | XP_011543150.1 | 7 | 1728 | |||
XM_017017359.1 | XP_016872848.1 | 7 | 1833 | |||
XM_017017360.1 | XP_016872849.1 | 7 | 1685 | |||
GenBank mRNA | AK298997.1 | - | 5 | 598 | ||
AK311690.1 | - | 7 | 866 | |||
AK313150.1 | - | 6 | 900 | |||
AK315892.1 | - | 3 | 294 | |||
BC012855.1 | - | 6 | 688 | 3863323 | ||
BI461207.1 | - | 6 | 659 | |||
DA004114.1 | - | 4 | 549 | |||
EU627066.1 | - | 6 | 688 | |||
EU627068.1 | - | 5 | 969 |