Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MARCH8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001002266.2 | NP_001002266.1 | 7 | 1059 | ||
NM_001282866.1 | NP_001269795.1 | 8 | 1905 | |||
NM_145021.5 | NP_659458.2 | 7 | 1547 | |||
XM_005271804.2 | XP_005271861.1 | 8 | 2394 | |||
XM_006717704.2 | XP_006717767.1 | 8 | 1809 | |||
XM_011539492.2 | XP_011537794.1 | 9 | 2445 | |||
XM_011539493.2 | XP_011537795.1 | 10 | 2686 | |||
XM_011539494.2 | XP_011537796.1 | 9 | 2635 | |||
XM_011539495.1 | XP_011537797.1 | 7 | 943 | |||
XM_017015894.1 | XP_016871383.1 | 8 | 1599 | |||
GenBank mRNA | AK023636.1 | - | 3 | 1348 | ||
AK057289.1 | - | 7 | 1253 | |||
AK307667.1 | - | 8 | 1905 | |||
AK313340.1 | - | 7 | 1016 | |||
AL833316.1 | - | 4 | 1290 | |||
BG433948.1 | - | 5 | 562 |