Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: EYA4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001301012.1 | NP_001287941.1 | 13, 14 | 1494 | ||
NM_001301013.1 | NP_001287942.1 | 15, 16 | 1674 | |||
NM_004100.4 | NP_004091.3 | 15, 16 | 1798 | |||
NM_172103.3 | NP_742101.2 | 14, 15 | 1729 | |||
NM_172105.3 | NP_742103.1 | 15, 16 | 1798 | |||
XM_005266851.4 | XP_005266908.1 | 15, 16 | 1523 | |||
XM_005266853.4 | XP_005266910.1 | 14, 15 | 1436 | |||
XM_017010368.1 | XP_016865857.1 | 16, 17 | 2043 | |||
XM_017010369.1 | XP_016865858.1 | 15, 16 | 1505 | |||
XM_017010370.1 | XP_016865859.1 | 14, 15 | 1454 | |||
XM_017010371.1 | XP_016865860.1 | 14, 15 | 1439 | |||
XM_017010372.1 | XP_016865861.1 | 13, 14 | 1361 | |||
XM_017010373.1 | XP_016865862.1 | 13, 14 | 1343 | |||
XM_017010374.1 | XP_016865863.1 | 13, 14 | 1660 | |||
XM_017010375.1 | XP_016865864.1 | 13, 14 | 1634 | |||
GenBank mRNA | AK295798.1 | - | 13, 14 | 1661 | ||
AK299378.1 | - | 16, 17 | 1588 | |||
AK301950.1 | - | 14, 15 | 1734 | |||
BC014193.2 | - | 7, 8 | 655 | |||
BC041063.1 | - | 14, 15 | 1668 | 5458911 | ||
KU178044.1 | - | 13, 14 | 1271 | |||
KU178045.1 | - | 12, 13 | 1178 | |||
Y17114.1 | - | 15, 16 | 1781 |