Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC39A12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001145195.1 | NP_001138667.1 | 5 | 1064 | ||
NM_001282733.1 | NP_001269662.1 | 5 | 1064 | |||
NM_001282734.1 | NP_001269663.1 | 4 | 717 | |||
NM_152725.3 | NP_689938.2 | 5 | 1064 | |||
GenBank mRNA | AK055061.1 | - | 5 | 1012 | ||
AK295294.1 | - | 4 | 593 | |||
AK303735.1 | - | 5 | 749 | |||
BC035118.1 | - | 5 | 993 | |||
BC047635.1 | - | 5 | 979 | 4820920 | ||
BC065917.1 | - | 4 | 808 | |||
BC094700.1 | - | 5 | 940 | 30407123 | ||
BC117323.1 | - | 4 | 814 | 40125874 | ||
BC143551.1 | - | 4 | 813 | 9052062 |