Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ALKBH3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_139178.3 | NP_631917.1 | 9 | 1172 | ||
GenBank mRNA | AB042029.1 | - | 9 | 1135 | ||
AK315266.1 | - | 9 | 1239 | |||
BC103812.1 | - | 9 | 910 | 40000372 |