Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CPNE2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_152727.5 | NP_689940.3 | 10, 11 | 1211 | ||
GenBank mRNA | AF492484.1 | - | 10, 11 | 1162 | ||
AK292269.1 | - | 9, 10 | 1120 | |||
AK315394.1 | - | 10, 11 | 1114 | |||
BC053658.1 | - | 11, 12 | 1285 | 5750192 | ||
CR749617.1 | - | 9, 10 | 872 |