Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NT5DC1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_152729.2 | NP_689942.2 | 5 | 529 | ||
XM_006715378.3 | XP_006715441.1 | 5 | 532 | |||
GenBank mRNA | AK090918.1 | - | 2 | 1259 | ||
AK290408.1 | - | 5 | 479 | |||
AK308337.1 | - | 5 | 512 | |||
AY203935.1 | - | 2 | 139 | |||
BC015138.1 | - | 4 | 309 | |||
BC108699.1 | - | 5 | 486 | 6061738 | ||
BC136832.1 | - | 5 | 488 | 9020823 | ||
BC136841.1 | - | 5 | 488 | 9020832 | ||
BM808222.1 | - | 518 |