Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ADGRF5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001098518.1 | NP_001091988.1 | 20, 21 | 4249 | ||
NM_015234.4 | NP_056049.4 | 20, 21 | 4189 | |||
XM_005248892.2 | XP_005248949.1 | 21, 22 | 4369 | |||
XM_005248893.3 | XP_005248950.1 | 21, 22 | 4325 | |||
XM_011514354.2 | XP_011512656.1 | 22, 23 | 4392 | |||
XM_017010422.1 | XP_016865911.1 | 20, 21 | 4165 | |||
XM_017010423.1 | XP_016865912.1 | 21, 22 | 4272 | |||
GenBank mRNA | AB018301.2 | - | 20, 21 | 4187 | ||
AB031421.1 | - | 19, 20 | 4004 | |||
AK075121.1 | - | 4, 5 | 1137 | |||
AK289503.1 | - | 20, 21 | 4187 | |||
AK300380.1 | - | 9, 10 | 2760 | |||
AL832125.1 | - | 18, 19 | 3730 | |||
AY498875.1 | - | 20, 21 | 4048 | |||
BC050087.1 | - | 7, 8 | 2073 | |||
BC066121.1 | - | 20, 21 | 4027 | 5298732 |