Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other ADGRF5 SIRNA Assays ›
Gene Symbol:
ADGRF5
Gene Name:
adhesion G protein-coupled receptor F5
Gene Aliases:
GPR116, KPG_001
Chromosome Location:
Chr.6: 46852505 - 46954954 on Build GRCh38
UniGene ID:
Hs.362806
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
s48032
Purification
HPLC
Standard
HPLC-IVR
Size
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
Availability
Inventoried
Catalog #
4392420
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: ADGRF5
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001098518.1
NP_001091988.1
16, 17
2677
NM_015234.4
NP_056049.4
16, 17
2617
XM_005248892.2
XP_005248949.1
17, 18
2797
XM_005248893.3
XP_005248950.1
17, 18
2753
XM_005248894.3
XP_005248951.1
17, 18
2797
XM_005248895.3
XP_005248952.1
17, 18
2797
XM_011514353.2
XP_011512655.1
16, 17
2677
XM_011514354.2
XP_011512656.1
18, 19
2820
XM_017010418.1
XP_016865907.1
17, 18
2753
XM_017010419.1
XP_016865908.1
16, 17
2633
XM_017010420.1
XP_016865909.1
16, 17
2677
XM_017010421.1
XP_016865910.1
16, 17
2633
XM_017010422.1
XP_016865911.1
16, 17
2593
XM_017010423.1
XP_016865912.1
17, 18
2700
XM_017010424.1
XP_016865913.1
17, 18
2700
GenBank mRNA
AB018301.2
-
16, 17
2615
AB031421.1
-
15, 16
2432
AK289503.1
-
16, 17
2615
AK296948.1
-
6, 7
1000
AK300380.1
-
5, 6
1188
AL832125.1
-
14, 15
2158
AY140958.1
-
15, 16
2385
AY498875.1
-
16, 17
2476
BC050087.1
-
3, 4
501
BC066121.1
-
16, 17
2455
5298732
Back To Top
More Information
Set Membership:
G-protein coupled receptor
Gene Ontology Categories:
Function(s)
Process(es)
energy reserve metabolic process
cell surface receptor signaling pathway
G-protein coupled receptor signaling pathway
glucose homeostasis
negative regulation of macrophage activation
surfactant homeostasis
fat cell differentiation
positive regulation of phospholipid biosynthetic process
G-protein coupled receptor activity
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-7487cff77c-nnwcq:80/10.239.162.212:80.
git-commit: c531a7a49d92b854cb004d8acd820c41186dcba6
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.16.2-2024.09.38-1.0