Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SPATS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145026.3 | NP_659463.1 | 6 | 723 | ||
GenBank mRNA | AK058171.1 | - | 5 | 969 | ||
BC100956.2 | - | 5 | 778 | 40004668 | ||
BC100957.2 | - | 5 | 778 | 40004669 | ||
BC100958.2 | - | 5 | 778 | 40004672 | ||
BC100959.2 | - | 6 | 872 | 40004675 | ||
BC110065.1 | - | 5 | 778 | 40004676 | ||
BC111414.1 | - | 5 | 778 | 40004677 |