Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FGD2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_173558.3 | NP_775829.2 | 5 | 724 | ||
XM_011514372.2 | XP_011512674.1 | 5 | 716 | |||
XM_011514373.2 | XP_011512675.1 | 5 | 716 | |||
XM_017010429.1 | XP_016865918.1 | 5 | 716 | |||
GenBank mRNA | AK092732.1 | - | 5 | 672 | ||
AK097981.1 | - | 1 | 610 | |||
AK131079.1 | - | 1 | 133 | |||
AK290419.1 | - | 6 | 717 | |||
AK301156.1 | - | 3 | 489 | |||
BC023645.2 | - | 5 | 724 | 5087501 | ||
BC053655.1 | - | 6 | 669 | 5755893 | ||
BX648164.1 | - | 5 | 631 |