Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PI16 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001199159.1 | NP_001186088.1 | 6 | 1025 | ||
NM_153370.2 | NP_699201.2 | 5 | 1130 | |||
XM_011514375.2 | XP_011512677.1 | 5 | 959 | |||
GenBank mRNA | AK075470.1 | - | 5 | 825 | ||
AK302193.1 | - | 4 | 633 | |||
AK303928.1 | - | 4 | 779 | |||
AK312785.1 | - | 5 | 823 | |||
AY358422.1 | - | 5 | 816 | |||
BC022399.1 | - | 5 | 824 | 4691115 | ||
BC035634.1 | - | 5 | 1130 | 5217121 |