Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RNF182 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001165032.1 | NP_001158504.1 | 4 | 674 | ||
NM_001165033.1 | NP_001158505.1 | 2 | 453 | |||
NM_001165034.1 | NP_001158506.1 | 3 | 561 | |||
NM_152737.3 | NP_689950.1 | 3 | 591 | |||
XM_017010448.1 | XP_016865937.1 | 4 | 611 | |||
XM_017010449.1 | XP_016865938.1 | 3 | 675 | |||
XM_017010450.1 | XP_016865939.1 | 2 | 659 | |||
GenBank mRNA | AK090576.1 | - | 2 | 410 | ||
AK315528.1 | - | 3 | 572 | |||
BC030666.1 | - | 3 | 491 | 5259219 | ||
BC050030.1 | - | 2 | 315 | 5297801 | ||
DB176198.1 | - | 2 | 453 |