Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GPC2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_152742.2 | NP_689955.1 | 2 | 462 | ||
GenBank mRNA | AK096281.1 | - | 2 | 400 | ||
AL834418.1 | - | 2 | 348 | |||
AY093588.1 | - | 2 | 409 | |||
BC027972.1 | - | 2 | 403 | 5240315 |