Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SDK1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001079653.1 | NP_001073121.1 | 15 | 2132 | ||
NM_152744.3 | NP_689957.3 | 41 | 5987 | |||
XM_011515188.1 | XP_011513490.1 | 35 | 4828 | |||
XM_011515189.1 | XP_011513491.1 | 35 | 4825 | |||
XM_017011837.1 | XP_016867326.1 | 35 | 4828 | |||
GenBank mRNA | AK055987.1 | - | 2 | 58 | ||
AK074077.1 | - | 8 | 927 | |||
AK074083.1 | - | 26 | 3623 | |||
AL833354.1 | - | 15 | 2129 | |||
AY310398.1 | - | 41 | 5987 |