Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CCZ1B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_198097.3 | NP_932765.1 | 7 | 687 | ||
GenBank mRNA | AK292326.1 | - | 7 | 655 | ||
AK296026.1 | - | 6 | 493 | |||
AL050215.1 | - | 5 | 329 | |||
BC010130.2 | - | 7 | 622 | 3953739 |
Gene Symbol: CCZ1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015622.5 | NP_056437.4 | 7 | 679 | ||
GenBank mRNA | AF151801.1 | - | 7 | 723 | ||
BC067810.1 | - | 7 | 689 | |||
BC132749.1 | - | 7 | 664 | 40146771 | ||
BC132751.1 | - | 7 | 664 | 40146773 | ||
BI546694.1 | - | 7 | 670 | |||
CN310516.1 | - | 4 | 371 |