Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MTURN | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_152793.2 | NP_690006.2 | 1 | 298 | ||
XM_005249652.3 | XP_005249709.1 | 1 | 241 | |||
GenBank mRNA | AK096533.1 | - | 1 | 241 | ||
AK294982.1 | - | 1 | 28 | |||
AL834391.1 | - | 1 | 232 | |||
BI560774.1 | - | 1 | 302 | |||
DT217816.1 | - | 1 | 233 |